missing translation for 'onlineSavingsMsg'
Learn More

EIF3A Mouse, anti-Human, CoraLite Plus 488, Clone: 1C7B4, Proteintech

Product Code. 18004305 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18004305 100 μL 100µL
18058215 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18004305 Supplier Proteintech Supplier No. CL48867713100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The EIF3A antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human EIF3A. This antibody recognizes human, mouse antigen. The EIF3A antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.

Specifications

Antigen EIF3A
Applications Immunofluorescence
Clone 1C7B4
Concentration 1000 μg/mL
Conjugate CoraLite Plus 488
Gene Symbols eIF 3 theta, EIF3, eIF3 p167, eIF3 p170, eIF3 p180, eIF3 p185, eIF3 theta, EIF3A, EIF3S10, KIAA0139, P167, p180, p185, TIF32
Host Species Mouse
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 8661
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Antibody Molecular Weight 155 kDa
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.