missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eIF2b gamma Monoclonal Antibody (1H3)
GREENER_CHOICE

Product Code. 13431007
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13431007 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13431007 Supplier Invitrogen™ Supplier No. MA1078

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA1-078 detects eIF2B3 in human, non-human primate and rat samples and has been successfully used in Western blot, ICC/IF, IHC (P) and immunoprecipitation applications. The immunogen for MA1-078 is full length human eIF2B3 produced in HEK293T cells.

EIF2B3 catalyzes the exchange of eukaryotic initiation factor 2-bound GDP for GTP. Mutations in each of the five subunits of translation initiation factor eIF2B, including subunit 3, can cause leukoencephalopathy with vanishing white matter. EIF2B3 has also been identified as a cofactor of hepatitis C virus internal ribosome entry site-mediated translation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF2b gamma
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, RNA Immunoprecipitation, Immunocytochemistry
Classification Monoclonal
Clone 1H3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide
Gene EIF2B3
Gene Accession No. P70541, Q9NR50
Gene Alias 1190002P15Rik; EIF-2B; eIF-2B GDP-GTP exchange factor subunit gamma; EIF2B3; Eif2bg; EIF2Bgamma; eukaryotic translation initiation factor 2B subunit gamma; eukaryotic translation initiation factor 2B, subunit 3; eukaryotic translation initiation factor 2B, subunit 3 (gamma, 58kD); eukaryotic translation initiation factor 2B, subunit 3 gamma; eukaryotic translation initiation factor 2B, subunit 3 gamma, 58kDa; translation initiation factor eIF-2B subunit gamma; Translation initiation factor eIF-2B subunit gamma-like protein
Gene Symbols EIF2B3
Host Species Mouse
Immunogen Full length human eIF2B3 produced in HEK293T cells.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171145, 8891
Target Species Human, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.