Learn More
EIF1AY Monoclonal Antibody (OTI2G2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA807511
Encompass_Preferred
 View more versions of this product
Product Code. 17568053
  • 540.87 EUR / 100µL
Description
This gene encodes a protein similar to eukaryotic translation initiation factor 1A (EIF1A). EIF1A is required for the binding of the 43S complex (a 40S subunit, eIF2/GTP/Met-tRNAi and eIF3) to the 5′ end of capped RNA.
Specifications
Specifications
EIF1AY
Monoclonal
Unconjugated
EIF1AY
eIF-4C
Mouse
Affinity Chromatography
RUO
9086
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
OTI2G2
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O14602
EIF1AY
Human recombit protein fragment corresponding to amino acids 2-144 of human EIF1AY produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2a
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions