Learn More
EIF1AY Monoclonal Antibody (OTI2G2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807511
This item is not returnable.
View return policy
Description
This gene encodes a protein similar to eukaryotic translation initiation factor 1A (EIF1A). EIF1A is required for the binding of the 43S complex (a 40S subunit, eIF2/GTP/Met-tRNAi and eIF3) to the 5′ end of capped RNA.Specifications
| EIF1AY | |
| Monoclonal | |
| Unconjugated | |
| EIF1AY | |
| eIF-4C | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9086 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI2G2 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O14602 | |
| EIF1AY | |
| Human recombit protein fragment corresponding to amino acids 2-144 of human EIF1AY produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |