missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF1A Antibody (SR1285), Novus Biologicals™
Rabbit Monoclonal Antibody
338.00 EUR - 449.00 EUR
Tekniske data
| Antigen | EIF1A |
|---|---|
| Clone | SR1285 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Produktkode | Brand | Quantity | Pris | Mængde & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Quantity | Pris | Mængde & tilgængelighed | |||||
|
18616464
|
Novus Biologicals
NBP3-22578-100ul |
100 μg |
449.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18640834
|
Novus Biologicals
NBP3-22578-50ul |
50 μg |
338.00 EUR
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivelse
EIF1A Monoclonal antibody specifically detects EIF1A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationTekniske data
| EIF1A | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
| 1964 | |
| IgG | |
| Affinity purified |
| SR1285 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human | |
| eIF-1A, EIF1AP1, eIF-4C, EIF4C, eukaryotic translation initiation factor 1A, X chromosome, eukaryotic translation initiation factor 1A, X-chromosomal, eukaryotic translation initiation factor 1A, Y chromosome, eukaryotic translation initiation factor 1A, Y-chromosomal, Eukaryotic translation initiation factor 4C, FLJ45762, Putative eukaryotic translation initiation factor 1A | |
| A synthesized peptide derived from human EIF1A (Uniprot #: P47813) | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel