missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGR4 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-06032
This item is not returnable.
View return policy
Description
EGR4 Polyclonal specifically detects EGR4 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| EGR4 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| AT133, early growth response 4, early growth response protein 4, EGR-4, NGFIC, NGFI-C, PAT133 | |
| Goat | |
| 51 kDa | |
| 0.1 mg | |
| Cell Cycle and Replication | |
| 1961 | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:4000 | |
| G3V1T5 | |
| EGR4 | |
| Peptide with sequence NCGSQGDYQAAPE corresponding to internal region according to NP_001956.2. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction