missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EGR1 Monoclonal Antibody (T.160.5)

Rabbit Monoclonal Antibody
Brand: Invitrogen™ MA515008
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
The protein encoded by this gene belongs to the EGR family of C2H2-type zinc-finger proteins. It is a nuclear protein and functions as a transcriptional regulator. The products of target genes it activates are required for differentitation and mitogenesis. Studies suggest this is a cancer suppresor gene.
Specifications
| EGR1 | |
| Monoclonal | |
| 42 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5 | |
| P08046, P08154, P18146 | |
| EGR1 | |
| Synthetic peptide corresponding to the amino terminus of the sequence of human EGR1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ChIP Assay, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| T.160.5 | |
| Unconjugated | |
| EGR1 | |
| A530045N19Rik; AT225; early growth response 1; early growth response protein 1; egr; EGR1; EGR-1; ETR103; G0S30; Krox-1; Krox24; Krox-24; Krox-24 nuclear protein; Nerve growth factor-induced protein A; Ngf1; NGF1-A; Ngfi; NGFIA; NGFI-A; TIS8; transcription factor ETR103; Transcription factor Zif268; Zenk; Zfp-6; Zif268; zif-268; Zinc finger protein 225; zinc finger protein Krox-24; ZNF225 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 13653, 1958, 24330 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction