missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EEA1 Monoclonal Antibody (F.43.1)

Rabbit Monoclonal Antibody
Brand: Invitrogen™ MA514794
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
Early endosomal antigen 1 (EEA1) is a 162 kDa membrane bound protein component specific to the early endosome and is essential for fusion between early endocytic vesicles. Early endosomes are cytoplasmic compartments which fuse with endocytic vesicles for redistribution of extracellular compounds to alternate destinations. Zinc-finger-like domains, reminiscent of those found in nucleic acid binding proteins, are located in the amino and carboxyl-terminal domains of EEA1. The carboxyl-terminal zinc-finger-like-domain is conserved in several other non-nuclear proteins, some of which are also involved in intracellular protein trafficking. In addition, this domain is an authentic zinc-binding domain and is critical to the intracellular localization of EEA1. Membrane association of EEA1 has been shown dependent on phosphatidyl 3-kinase activity, inhibitors of which cause EEA1 to dissociate from early endosomes.
Specifications
| EEA1 | |
| Monoclonal | |
| 44 μg/mL | |
| 0.01M HEPES with 0.15M NaCl, 100 μg/mL BSA, 50% glycerol and <0.02% sodium azide, pH 7.5 | |
| Q15075, Q8BL66 | |
| EEA1 | |
| Synthetic peptide corresponding to residues surrounding Ser70 of human EEA1 protein. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| F.43.1 | |
| Unconjugated | |
| EEA1 | |
| A430109M19Rik; B230358H09Rik; Early endosome antigen 1; early endosome antigen 1, 162kD; early endosome-associated protein; EEA1; Endosome-associated protein p162; MST105; MSTP105; ZFYVE2; Zinc finger FYVE domain-containing protein 2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 216238, 314764, 8411 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction