missing translation for 'onlineSavingsMsg'
Learn More

EDTA Disodium Salt Dihydrate, ACS Reagent Grade, Scharlab

Product Code. 11324104
Change view
Click to view available options
Quantity:
1 kg
25 kg
Unit Size:
1kg
25kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11324104 1 kg 1kg
11334104 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11324104 Supplier Scharlab Supplier No. AC09651000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as a sequestering agent in analytical chemistry.

  • ACS Reagent Grade
  • Synonyms: Ethylenediaminetetraacetic acid disodium salt dehydrate, Edetic acid disodium salt, Disodium dihydrogen ethylenediaminetetraacetate
  • Molecular Formula: C10H14N2Na2O8 • 2H2O
  • Molar Mass: 372.24g/mol
  • CAS: 6381-92-6
  • EIN ECS No.: 205-358-3
  • Loss on Drying: 8.7–11.4% (150°C, 6h)
  • Residue on Ignition: 0.2% max

Chemical Identifiers

CAS 6381-92-6
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
MDL Number MFCD00150037,MFCD00003541
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
Synonym edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
PubChem CID 44120005
IUPAC Name disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

Melting Point 252°C (decomposition)
Color White
pH 4.0 to 5.0 (5%, H2O, 20°C)
Quantity 1 kg
Solubility Information 100g/L at 20°C Solubility in water
Formula Weight 372.24g/mol
Percent Purity 99 to 101% (complexometric)
Grade ACS Reagent
Physical Form Crystalline Powder
compliance-icons
Product Identifier
  • Ethylenediaminetetraacetic acid, EDTA, disodium salt, dihydrate, reagent grade, ACS, Reag. Ph Eur
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.