missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDTA diNA Salt 0.005 M For Laboratory Use and TH Titrations, Chem Lab
Click to view available options
Quantity:
5 L
Unit Size:
5 L
Specifications
Specifications
| Color | Colorless |
| CAS | 6381-92-6 |
| Boiling Point | 100°C |
| Melting Point | 0°C |
| Concentration | 0.005 M |
| Density | 1 g/mL |
| Molecular Weight (g/mol) | 372.24 |
| Packaging | Polyethylene |
| Traceability to NIST | Reference standards acc. NIST SRM 915b |
| Quantity | 5 L |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction