missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDTA Concentrated Volumetric Solution, To Make 1 L of 0.05 M (0.1 N) EDTA (Disodium Salt), Reagecon
Shop All Reagecon Products

Description
- Tested to ± 0.2%
- May be used for manual titrations or more commonly automatic titrations
- Must be diluted accurately to 1 L in a volumetric flask under controlled conditions for optimal performance
Specifications
Specifications
| Chemical Name or Material | EDTA Concentrated Volumetric Solution |
| Packaging | Ampoule |
| Physical Form | Liquid |
| Technique | Titration |
| UN Number | 1824 |
| Quantity | 1 Ampule |
Safety and Handling
Signal Word
- Danger
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- H314-Causes severe skin burns and eye damage.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: EDTA, sodium hydroxide
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?