Learn More
EDTA Analytical Volumetric Di Sodium Solution, Reagecon™
Shop All Reagecon Products
Click to view available options
Quantity:
10 L
Unit Size:
10 L
Specifications
Specifications
| Quantity | 10 L |
| Concentration or Composition (by Analyte or Components) | 0.01M |
| Physical Form | Liquid |