Description
This Bel-Art™ paperboard spoon stores flat and easily folds into a functional sampling tool for retrieving materials in the laboratory.
- Plastic free paper spoon is 100% compostable and recyclable
- Uses 50% to 90% less material and space than plastic laboratory spoons
- Low cost way to be eco-friendly in the laboratory
- Can be used as a weigh boat eliminating the need for glassine sheets
Made in the USA
NOTE: EcoTensil™ Spoons are not meant for measuring exact quantities of materials.
Specifications
Specifications
| Sterility | Non-sterile |
| Autoclavable | Not Autoclavable |
| Capacity (Metric) | 10 mL |
| Disposable | Limited-use |
| Product Type | Earth Friendly Sampling Spoon |
| Dimensions (L x W) | 5 x 1.5 in. (12.7 x 3.8cm) |
| Material | Paperboard |