Learn More
Econazole EP Impurity B Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Econazole EP Impurity B Hydrochloride |
| CAS | 2334059-11-7 |
| Molecular Formula | C15H14Cl3NO . HCl |
| Quantity | 10 mg |
| Synonym | 2-((4-chlorobenzyl)oxy)-2-(2,4-dichlorophenyl)ethan-1-amine, hydrochloride (1:1) |
| InChI Formula | InChI=1S/C15H14Cl3NO/c16-11-3-1-10(2-4-11)9-20-15(8-19)13-6-5-12(17)7-14(13)18/h1-7,15H,8-9,19H2 |
| SMILES | ClC1=CC=C(COC(CN)C2=CC=C(C=C2Cl)Cl)C=C1.Cl |
| Molecular Weight (g/mol) | 330.6 |