Learn More
Sharpsguard™ Eco EXTRA 8.5 Waste Container
Click to view available options
Color:
Orange
Yellow
Unit Size:
Pack of 15
Description
- Sharpsguard™ eco EXTRA 8.5 is an ideal solution for the management of sharps in high public access areas.
- This container is supplied with the base and lid separately. Details about how to lock the container and keep the users and contents safe can be found on the label.
- This container is for disposal by incineration or other authorized process and is designed for the consignment of waste categorized by EWC codes: 18-01-01, 18-01-03, 18-01-09, 18-02-01, 18-02-02, 18-02-08.
Specifications
Specifications
| Capacity (Metric) | 7.59 L |
| Color | Yellow |
| Dimensions (L x W x H) | 360 x 161 x 269 mm |
| Quantity | 15/Pk. |
| Weight (Metric) | 816 g |