Learn More
Sharpsguard™ Eco EXTRA 8.5 Waste Container
Product Code. 17744864
Click to view available options
Color:
Orange
Yellow
Unit Size:
Pack of 15
Product Code. 17744864
Brand: Sharpsguard™ DD580OLR
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The new Sharpsguard™ eco EXTRA 8.5 container, made using at least 40% recycled plastic that is used for the disposal of sharps that have been contaminated with medicines, but not cytotoxic.
  • Sharpsguard™ eco EXTRA 8.5 is an ideal solution for the management of sharps in high public access areas.
  • This container is supplied with the base and lid separately. Details about how to lock the container and keep the users and contents safe can be found on the label.
  • This container is for disposal by incineration or other authorized process and is designed for the consignment of waste categorized by EWC codes: 18-01-01, 18-01-03, 18-01-09, 18-02-01, 18-02-02, 18-02-08.
Specifications
Capacity (Metric) 7.59 L
Color Orange
Dimensions (L x W x H) 360 x 161 x 269 mm
Quantity 15/Pk.
Weight (Metric) 816 g