Learn More
Sharpsguard™ Eco Cyto 22 Waste Container
Click to view available options
Capacity (Metric):
17.86 L
Unit Size:
Each
Description
- Sharpsguard™ eco cyto 22 is ideal for theaters, laboratories and sluice rooms with high volumes of cytotoxic and cytostatic waste. The wide aperture allows economical and convenient disposal of large items.
- This container is supplied with the base and lid separately and contains an absorbent pad. Details about how to lock the container and keep the users and contents safe can be found on the label.
- It is designed to fit safely and securely into our POUDS™ mobile holders and bracket range.
- This container is for disposal by incineration or other authorized process and is designed for the consignment of waste categorized by EWC codes: 18-01-01, 18-01-03, 18-01-08, 18-01-09, 18-02-01, 18-02-02, 18-02-07, 18-02-08.
Specifications
Specifications
| Capacity (Metric) | 17.86 L |
| Closure Color | Purple |
| Diameter (Metric) Outer | 327 mm |
| Color | Yellow |
| Height (Metric) | 343 mm |
| Quantity | 1 Ea. |
| Weight (Metric) | 943 g |