Learn More
Sharpsguard™ Eco Anatomical Waste Container
GREENER_CHOICE
Product Code. 17764874
Change view
Click to view available options
Capacity (Metric):
2.6 L
10.91 L
21.01 L
Unit Size:
Pack of 10
Pack of 20
Pack of 48
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
17764874 2.6 L Pack of 48
17784834 21.01 L Pack of 10
17754874 10.91 L Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17764874 Supplier Sharpsguard™ Supplier No. DD912R
The new Sharpsguard™ eco anatomical container, made using at least 40% recycled plastic with red solid lid is for the disposal of anatomical waste and other potentially infectious waste.
  • Sharpsguard™ eco anatomical is for the safe disposal of anatomical and other potentially infectious waste.
Specifications
Capacity (Metric) 2.6 L
Closure Color Red
Diameter (Metric) Outer 178 mm
Color Yellow
Height (Metric) 166 mm
For Use With (Application) Safe Disposal of Anatomical and Potentially Infectious Waste