Learn More
Elma™ EC 35 Cleaner
Product Code. 10018191
Click to view available options
Volume (Metric):
1 L
Unit Size:
1 L
Product Code. 10018191
Brand: Elma™ 5814100000
This highly effective cleaner with active oxygen easily removes organic contaminants, deposits and food residues in an ultrasonic bath.
  • Suitable for dental prostheses made of metal, ceramics and plastics.
  • This highly effective celaner with active oxygen easily removes organic contaminants, deposits and food residues in an ultrasonic bath
Specifications
Product Type EC 35 Cleaner
Form Liquid
pH 4 to 5
Appearance Blue
Boiling Point 100°C
Certifications/Compliance CE
Container Type Bottle
Content And Storage Above -5°C to 35°C
For Use With (Application) For clean dental prostheses
Volume (Metric) 1 L
Product Identifier
  • Cleaning solution ELMA-CLEAN 35, for dental, 1L .
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P273-Avoid release to the environment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Sulphuric acid, propan-2-ol, peracetic acid