missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBOV VP40 protein Antibody (02), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-12761V
This item is not returnable.
View return policy
Description
EBOV VP40 protein Monoclonal antibody specifically detects EBOV VP40 protein in Ebola virus samples. It is validated for Western Blot, ELISA
Specifications
| EBOV VP40 protein | |
| Monoclonal | |
| DyLight 405 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Ebola virus | |
| Purified |
| Western Blot, ELISA | |
| 02 | |
| 50mM Sodium Borate | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant EBOV VP40 protein (Accession#: YP_138522.1; Val31-Lys326). | |
| 100 μg | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction