Learn More
STEMCELL Technologies EasySep™ Multistand with EasySep™ Magnets
Product Code. 17177841
Encompass_Preferred
Click to view available options
Includes:
4 Magnets
Unit Size:
1 set
Product Code. 17177841
Brand: STEMCELL Technologies 18004
Immunomagnetic column-free magnet
  • The EasySep™ Multistand allows the separation of up to 4 samples at one time
  • An ingenious reversible clamping bar allows the Multistand to hold up to 4 EasySep™ (Catalog #18000) or 4 'The Big Easy' EasySep™ Magnets (Catalog #18001)
  • After each round of magnetic separation, the supernatants from all samples are poured off simultaneously into the receiving tubes
  • The receiving tube bar is indexed to ensure proper position of the tubes
Specifications
Includes 4 Magnets
For Use With (Application) Cell Isolation
Quantity 1 Ea.