Learn More
Greiner Bio-One EASYseal™ Adhesive Microplate Sealer
Product Code. 15975331
Click to view available options
:
EASYseal Adhesive Microplate Sealer
Product Code. 15975331
Brand: Greiner Bio-One 676001
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For optical measurements and all laboratory purposes. Greiner Bio-One EASYseal™ Adhesive Microplate Sealer provides protection against evaporation and contamination.
  • Coated with acrylate adhesive
  • Coverage/storage of microplates (e.g in ELISA applications)
  • Protection against evaporation and contamination
  • Suitable for optical measurements
TRUSTED_SUSTAINABILITY
Specifications
Color Transparent
Sterility Non-sterile
No. per Case 100
For Use With (Application) ELISA
Product Type Sealer
Adhesive Acrylate
Plate Compatibility Microplate
For research use only. Not for use in diagnostic procedures, unless otherwise indicated.