missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One EASYseal™ Adhesive Microplate Sealer

Product Code. 13254833
Click to view available options
:
EASYseal Adhesive Microplate Sealer
This item is not returnable. View return policy

Product Code. 13254833

Brand: Greiner Bio-One 676001

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For optical measurements and all laboratory purposes. Greiner Bio-One EASYseal™ Adhesive Microplate Sealer provides protection against evaporation and contamination.

  • Coated with acrylate adhesive
  • Coverage/storage of microplates (e.g in ELISA applications)
  • Protection against evaporation and contamination
  • Suitable for optical measurements
TRUSTED_SUSTAINABILITY

Specifications

Color Transparent
Sterility Non-sterile
No. per Case 100
For Use With (Application) ELISA
Product Type Sealer
Adhesive Acrylate
Plate Compatibility Microplate

For research use only. Not for use in diagnostic procedures, unless otherwise indicated.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.