missing translation for 'onlineSavingsMsg'
Learn More

EAF2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17121164 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17121164 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17121164 Supplier Thermo Scientific Supplier No. PA5113301

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

EAF2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen EAF2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.85 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Eaf2
Gene Accession No. Q811X5, Q91ZD6, Q96CJ1
Gene Alias AW048865; BM040; BM-040; Eaf2; Ehrlich S-II transcriptional activator factor; ELL associated factor 2; ELL-associated factor 2; Festa; FESTA-L; FESTA-S; testosterone regulated apoptosis inducer and tumor suppressor; testosterone-regulated apoptosis inducer and tumor suppressor protein; Traits; U19
Gene Symbols Eaf2
Host Species Rabbit
Immunogen Recombinant Human ELL-associated factor 2 protein (1-170AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 106389, 266787, 55840
Target Species Human, Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.