missing translation for 'onlineSavingsMsg'
Learn More

Solveco EA-50 Pap 2A Reagent

Product Code. 18032183
Click to view available options
Quantity:
1 L
Unit Size:
1 L
This item is not returnable. View return policy

Product Code. 18032183

Brand: Solveco EA50OT1L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This one-step polychromatic reagent is specifically designed as a counterstain for gynecological samples in cytology. It facilitates rapid staining and allows for clear differentiation between epithelial cells, with their cytoplasm appearing blue and pink.

OG-EA50 Pap reagent is an alcoholic polychromatic solution designed for use in the Papanicolaou staining method. The first step involves staining the nuclei with a hematoxylin solution, followed by counterstaining. Unlike traditional methods that require three steps, the OG-EA50 Pap reagent streamlines this process into just two steps, as it combines the OG-6 and EA50 reagents. The Orange G component stains the cytoplasm, remaining only in mature, keratinized cells at later stages. The polychromatic EA50 solution targets the non-stained areas of cells, effectively staining squamous cells, nucleoli, cilia, and erythrocytes. This reagent is suitable for both gynecological and non-gynecological samples, including sputum, urine, and cytological puncture specimens. For optimal staining results, OG-EA50 Pap reagent is fully compatible with other BioGnost reagents for cytological smears according to Papanicolaou, such as Hematoxylin HP, Pap 1A, and Hematoxylin HP, Pap 1B reagents.

Specifications

Product Type Reagent
Quantity 1 L
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1L EA-50 Pap 2A
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H371-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P370+P378-In case of fire: Use water mist/carbon dioxide/alcohol-resistant foam to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol, methanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.