missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E2F7 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24489-1-AP-20UL
This item is not returnable.
View return policy
Description
E2F transcription factors, such as E2F7, play an essential role in the regulation of cell cycle progression.Specifications
| E2F7 | |
| Polyclonal | |
| Unconjugated | |
| E2F7 | |
| E2F transcription factor 7, E2F7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 144455 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.38 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q96AV8 | |
| E2F7 | |
| E2F7 Fusion Protein Ag21599 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction