missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E2F-4 Antibody (S09-7E6), Novus Biologicals™
Rabbit Monoclonal Antibody
421.00 EUR - 645.00 EUR
Specifications
| Antigen | E2F-4 |
|---|---|
| Clone | S09-7E6 |
| Concentration | 0.3 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18601545
|
Novus Biologicals
NBP3-19623-50ul |
50 μg |
421.00 EUR
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18663054
|
Novus Biologicals
NBP3-19623-100ul |
100 μg |
645.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
E2F-4 Monoclonal antibody specifically detects E2F-4 in Human samples. It is validated for Western BlotSpecifications
| E2F-4 | |
| 0.3 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| E2F transcription factor 4, p107/p130-binding, E2F-4p107/p130-binding protein, transcription factor E2F4 | |
| A synthetic peptide of human E2F-4 (Uniprot # Q16254) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| S09-7E6 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 1874 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title