Learn More
(E,Z)-Epalrestat Methyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | (E,Z)-Epalrestat Methyl Ester |
| CAS | 682775-70-8 |
| Molecular Formula | C16 H15 N O3 S2 |
| Quantity | 25 mg |
| Synonym | (E,Z)-5-(2-Methyl-3-phenyl-2-propen-1-ylidene)-4-oxo-2-thioxo-3-thiazolidineacetic Acid Methyl Ester |
| InChI Formula | InChI=1S/C16H15NO3S2/c1-11(8-12-6-4-3-5-7-12)9-13-15(19)17(16(21)22-13)10-14(18)20-2/h3-9H,10H2,1-2H3 |
| SMILES | COC(=O)CN1C(=S)SC(=CC(=Cc2ccccc2)C)C1=O |
| IUPAC Name | methyl 2-[5-(2-methyl-3-phenyl-prop-2-enylidene)-4-oxo-2-thioxo-thiazolidin-3-yl]acetate |
| Molecular Weight (g/mol) | 333.43 |
| Formula Weight | 333.0493 |