missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(E)-LHF-535, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
(E)-LHF-535 is the E-isomer of LHF-535. LHF-535 is an antiviral agent extracted from patent WO2013123215A2, Compound 38, has EC50s of <1 μM, <1 μM, <1 μM, and 1-10 μM for Lassa, Machupo, Junin, and VSVg virus, respectively.
Specifications
Specifications
| Chemical Name or Material | (E)-LHF-535 |
| Physical Form | Solid |
| Color | Light Yellow |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C27H28N2O2 |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Solubility Information | DMSO : 60 mg/mL (145.45 mM; Need ultrasonic) |
| SMILES | CC(O)(C1=CC=C(/C=C/C2=CC=C3N(C4=CC=C(OC(C)C)C=C4)C=NC3=C2)C=C1)C |
| Molecular Weight (g/mol) | 412.52 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction