Learn More
Invitrogen™ E-Gel™ Agarose Gels Starter Pak, 2% (EU only)
Product Code. 10388982
Click to view available options
Quantity:
Each
Unit Size:
Each
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10388982
Brand: Invitrogen™ G600002EU
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
E-Gel™ Agarose Gels Starter Pak, 2% (EU only)
E-Gel™ Single Comb Gels are supplied in a 12-well mini-gel format, with a choice of either SYBR Safe™, or ethidium bromide DNA stain. Each gel contains a single row of 12 sample wells. E-Gels are available in two pack sizes-a Starter Pak, and 18-gel multi-paks.
TRUSTED_SUSTAINABILITY
Specifications
Description E-Gel™ Agarose Gels Starter Pak, 2%, E-Gel™ Gel Type, Ethidium Bromide In-Gel Dye, 12 Wells, Single Comb Well Format, Store at Room Temperature, For Nucleic Acid Gel Electrophoresis and Blotting
Concentration 2%
Product Type Agarose Gels Starter Kit
For Use With (Application) DNA and RNA purification analysis, nucleic acid gel electrophoresis and blotting, agarose gel electrophoresis
Kit Contents 6 E-Gels, the E-Gel™ PowerBase™ v.4, and a manual, 18-packs contain 18 gels and a manual
Quantity Each
Storage Requirements Room temperature