Learn More
Dysprosium AAS Standard, 1000 mg/L Dy in HNO3 2%, Honeywell Fluka™
Product Code. 16644304 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16644304 500 mL 500mL
16634304 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16644304 Supplier Honeywell Fluka Supplier No. E4024500ML
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Dysprosium AAS Standard
Density 1.01 g/cm3
pH <2
Quantity 500 mL