Learn More
Thermo Scientific™ DyNAzyme I & II DNA Polymerases
Get improved thermal stability at high temperatures with these Taq-like enzymes, Thermo Scientific™ DyNAzyme™ DNA Polymerases.
Brand: Thermo Scientific™ F501S
Additional Details : Weight : 0.01000kg
Description
DyNAzyme DNA Polymerases are Taq-like enzymes with improved thermal stability at high temperatures. They are ideal for all routine PCR applications, and have improved tolerance against DMSO. DyNAzyme I DNA Polymerase is a native thermostable DNA polymerase from Thermus brockianus. DyNAzyme II DNA Polymerase is an easy-to-use, recombinant thermostable DNA polymerase for standard PCR.
Highlights
- Improved thermal stability
- Improved tolerance against DMSO
- Supplied with optimized buffers or with Mg2+-free buffers and MgCl2 solution
- Detergent-free buffer also available
Specifications
DyNAzyme II DNA Polymerase, 10x Optimized DyNAzyme Buffer. Recommended storage temperature -20 °C. Stable for one year from the assay date. | |
DyNAzyme™ II DNA Polymerase | |
250U (2U/µL) | |
PCR |