Learn More
Thermo Scientific™ DyNAzyme I & II DNA Polymerases
Get improved thermal stability at high temperatures with these Taq-like enzymes, Thermo Scientific™ DyNAzyme™ DNA Polymerases.
70.50 EUR - 274.00 EUR
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10544219
|
Thermo Scientific™
F501L |
1000U (2U/µL) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10266269
|
Thermo Scientific™
F501S |
250U (2U/µL) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10104529
|
Thermo Scientific™
F503L |
1000U (2U/µL) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
DyNAzyme DNA Polymerases are Taq-like enzymes with improved thermal stability at high temperatures. They are ideal for all routine PCR applications, and have improved tolerance against DMSO. DyNAzyme I DNA Polymerase is a native thermostable DNA polymerase from Thermus brockianus. DyNAzyme II DNA Polymerase is an easy-to-use, recombinant thermostable DNA polymerase for standard PCR.
Highlights
- Improved thermal stability
- Improved tolerance against DMSO
- Supplied with optimized buffers or with Mg2+-free buffers and MgCl2 solution
- Detergent-free buffer also available