missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dynamin 2 Antibody (3F5F3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
203.00 EUR - 552.00 EUR
Specifications
| Antigen | Dynamin 2 |
|---|---|
| Clone | 3F5F3 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18272954
|
Novus Biologicals
NBP2-61757 |
0.1 mL |
552.00 EUR
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18673048
|
Novus Biologicals
NBP2-61757-0.025ml |
0.025 mL |
203.00 EUR
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dynamin 2 Monoclonal specifically detects Dynamin 2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
| Dynamin 2 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA | |
| Unconjugated | |
| Mouse | |
| CMTDI1CMT2M, CMTDIBdynamin II, DI-CMTB, DYN2EC 3.6.5.5, dynamin 2, dynamin-2, DYNII | |
| DNM2 | |
| IgG2a | |
| Protein G purified |
| 3F5F3 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
| Monoclonal | |
| Purified | |
| Apoptosis, Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neuroscience | |
| 1785 | |
| Purified recombinant fragment of human Dynamin-2 (AA: 520-744) expressed in E. Coli. | |
| Primary | |
| 98 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title