missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dynamin 2 Antibody (3F5C7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61758
This item is not returnable.
View return policy
Description
Dynamin 2 Monoclonal specifically detects Dynamin 2 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Dynamin 2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| CMTDI1CMT2M, CMTDIBdynamin II, DI-CMTB, DYN2EC 3.6.5.5, dynamin 2, dynamin-2, DYNII | |
| Mouse | |
| 98 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 3F5C7 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| DNM2 | |
| Purified recombinant fragment of human Dynamin-2 (AA: 520-744) expressed in E. Coli. | |
| Protein G purified | |
| Apoptosis, Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neuroscience | |
| 1785 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction