Learn More
Cole Parmer Dynalon Cuvette Stand
Shop All Cole Parmer Products
Click to view available options
Material:
Polyethylene
Unit Size:
Pack of 5
Description
Dynalon Cuvette Stand holds up to 12 standard-sized cuvettes
Accepts twelve 10-mm pathlength square cuvettes without obstructing tops. Polyethylene holder also stores cuvettes horizontally after use.
Specifications
Specifications
| Sterility | Non-sterile |
| Type | Cuvette Stand |
| Disposable | Reusable |
| Material | Polyethylene |
| For Use With (Equipment) | Standard Sized Cuvettes |
| For Use With (Application) | Holding 12 Standard Size Cuvettes |
| Shape | Rectangle |