missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynactin 2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16598931
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16598931 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16598931 Supplier Invitrogen™ Supplier No. PA596095

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: HIIVNPNAAY DKFKDKRVGT KGLDFSDRIG KTKRTGYESG EYEMLGEGLG VKETPQQKYQ RLLHEVQELT TEVEKIKTTV KESATEEKLT PVLLAKQLAA LKQQLVASHL EKLLGPDAAI NLTDPDGALA KRLLLQLEAT KNSKGGSGGK TTGTPPDSSL VTYELHSRPE QDKFSQAAKV AELEKRLTEL ETAVRCDQDA QNPLSAGLQG ACLMETVELL QAKVSALDLA VLDQVEARLQ SVLGKVNEIA KHKASVEDAD TQSKVHQLYE T; Positive Samples: MCF7; Cellular Location: Cytoplasm, Membrane, Peripheral membrane protein, centrosome, cytoskeleton, microtubule organizing center.

This gene encodes a 50-kD subunit of dynactin, a macromolecular complex consisting of 10-11 subunits ranging in size from 22 to 150 kDa. Dynactin binds to both microtubules and cytoplasmic dynein. It is involved in a diverse array of cellular functions, including ER-to-Golgi transport, the centripetal movement of lysosomes and endosomes, spindle formation, chromosome movement, nuclear positioning, and axonogenesis. This subunit is present in 4-5 copies per dynactin molecule. It contains three short alpha-helical coiled-coil domains that may mediate association with self or other dynactin subunits. It may interact directly with the largest subunit (p150) of dynactin and may affix p150 in place.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dynactin 2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DCTN2
Gene Accession No. Q13561, Q6AYH5
Gene Alias 2310042E05Rik; 50 kDa dynein-associated polypeptide; C130077D06Rik; C87049; DCTN2; dctn2 protein; DCTN50; DCTN-50; dynactin 2; dynactin 2 (p50); dynactin complex 50 kDa subunit; Dynactin subunit 2; dynamitin; epididymis secretory protein Li 77; GMP23-48K; Growth cone membrane protein 23-48K; HEL-S-77; hypothetical protein LOC527201; p50; p50 dynamitin; p50/dynamitin; RBP50; zgc:63867
Gene Symbols DCTN2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 50-320 of human DCTN2 (NP_0063911).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10540, 299850
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.