missing translation for 'onlineSavingsMsg'
Learn More

Dxd, MedChemExpress

Product Code. 16495778
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495778 10 mg 10mg
16421369 1 mg 1mg
16418457 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16495778 Supplier Medchem Express Supplier No. HY13631D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dxd (Exatecan derivative for ADC) is a potent DNA topoisomerase I inhibitor, with an IC50 of 0.31 μM, used as a conjugated drug of HER2-targeting ADC (DS-8201a).

Dxd (Exatecan derivative for ADC) is a potent DNA topoisomerase I inhibitor, with an IC50 of 0.31 μM, used as a conjugated drug of HER2-targeting ADC (DS-8201a).

Specifications

Chemical Name or Material Dxd
CAS 1599440-33-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C26H24FN3O6
Quantity 10 mg
Synonym Exatecan derivative for ADC
Solubility Information DMSO : 7.14 mg/mL (14.47 mM; Need ultrasonic)
SMILES O=C1[C@](O)(CC)C2=C(CO1)C(N3CC4=C5C6=C(CC[C@@H]5NC(CO)=O)C(C)=C(F)C=C6N=C4C3=C2)=O
Molecular Weight (g/mol) 493.48
Formula Weight 493.48
Percent Purity 98.02%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.