Learn More
DWK Life Sciences™ DURAN™ Vigreux Column, with 2 standard ground joints, complete, with slide-on glass jacket
Shop All DWK Life Sciences Products
Click to view available options
Standard Taper Joint:
NS 24/29
NS 29/32
Length (Metric):
450 mm
650 mm
Unit Size:
Each
Description
Vigreux columns are used in fractionated distillation for improved separation of substances. The slightly downward angled indentations considerably increase the contact surface between the gas phase and the glass, i.e. the cooling surface.
- Tight closure due to standard ground joint
- With slide-on glass jacket
Specifications
Specifications
| For Use With (Application) | Fractional distillation; Laboratory coolers, condensation of gases and vapors |
| Length (Metric) | 450 mm |
| Material | Boro 3.3 |
| Quantity | 1 Ea. |
| Standard Taper Joint | NS 29/32 |
| Autoclavable | Autoclavable |