Learn More
DWK Life Sciences DURAN™ Measuring Cylinder, without base, tall form, with spout, flat bottom, without graduation
Product Code. 31781800 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
Pack of 10
Pack of 50
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
31781800 100 mL -
15221719 250 mL Pack of 10
15231719 500 mL Pack of 10
15894498 25 mL Pack of 50
31781794 50 mL -
10420113 1000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 31781800 Supplier DWK Life Sciences Supplier No. 213932405
Only null left
Add to basket
Add to basket
For adjustment and printing.
DURAN™ measuring cylinders without base have an even wall thickness distribution over the entire measuring range, so that wedge errors are avoided. A plastic base is required for completion.
  • Glass articles uncalibrated (without graduation)
  • High shape, with spout and flat bottom
  • Uniform wall thickness distribution to avoid wedge errors
  • A separate plastic foot is required for completion.
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass (3.3)
Capacity (Metric) 100 mL
For Use With (Application) For adjustment and printing.; Mixing of several components
Includes 20/Pk.
Autoclavable Autoclavable