Learn More
Lenz Laborglasinstrumente™ Duran™ Glass GL 14 Thread Dimroth Condensers
Product Code. 10030182
Click to view available options
Standard Taper Joint No:
14/23
19/26
29/32
Length (Metric) Jacket:
160 mm
250 mm
400 mm
Unit Size:
Each
Product Code. 10030182
Brand: Lenz Laborglasinstrumente™ 05240314
Only null left
Specifications
Certifications/Compliance DIN 12591 and in addition to DIN standard
Description Cooling Surface: 440cm2
For Use With (Application) For condensing vapors
Length (Metric) Jacket 250 mm
Quantity 1 Each
Standard Taper Joint No 29/32
Material Duran Glass
Type Dimroth Condenser