Learn More
Lenz Laborglasinstrumente™ Duran Glass Columns acc. to Hempel
missing translation for 'orderingAttributeHoverText'
Length (Metric):
200mm
300mm
500mm
Includes:
Fixed Vacuum Jacket
NS Socket and Cone
NS Socket and Cone, Removable Glass jacket
missing translation for 'unitSize'
Each
Specifications
Specifications
| Includes | NS Socket and Cone, Removable Glass jacket |
| Length (Metric) | 300mm |
| Type | Columns acc. to Hempel |
| Material | Duran Glass |
| Quantity | 1Each |