Learn More
DWK Life Sciences DURAN™ GL 45 Aluminum Screw Closure, Silver
Shop All DWK Life Sciences Products
Click to view available options
Product Type:
Open Top Screw Cap
Screw Cap
Unit Size:
Pack of 10
Description
Aluminum is an alternative material to the plastic used in most screw closures for laboratory bottles. The caps are available as a closed-top cap with a high temperature PTFE coated cap liner, or as an open-top cap for use with septum or stopper. Metal closures offer a wide range of temperature tolerances and have a high mechanical strength. Aluminum offers superior barrier qualities that protect against light, moisture, oxygen, and other gases.
- Free of hydrocarbon-based plastics, plasticizers, phthalates, etc.
- High temperature usage up to +260 °C for depyrogensation or hot air sterilization
- Convenient: Thread fits all DURAN™ GL 45 glass bottles from 100 mL to 20 Litres
- Secure sealing with USP Class VI PTFE coated silicone cap liner
- Cap liner is supplied unmounted for easy processing and is replaceable
- Convenient opening and re-closing as the knurling provides extra grip for wet or gloved hands
- Protection: Electrically conductive material – ESD dissipation
Specifications
Specifications
| Color | Silver |
| For Use With (Application) | An Alternative to Plastic Screw Caps for Laboratory Bottles; High-Temperature Processing and Applications; Low-Temperature / Cryogenic Sample Storage; Pharma API and Sensitive Material Storage |
| Material | Aluminum |
| Quantity | 10/Pk. |
| Thread Size (Metric) | GL 45 |
| Liner Material | PTFE-coated Silicone |
| Height (Metric) | 25 mm |
| Product Type | Screw Cap |
| Diameter (Metric) | 50 mm |