Learn More
Si Analytics™ DURAN™ Burette Cylinder
Shop All SI Analytics Products
Click to view available options
Capacity (Metric):
10 mL
50 mL
Unit Size:
Each
Description
- Burette cylinder
- For use wtih interchangable units
Specifications
Specifications
| Color | Colorless |
| Capacity (Metric) | 10 mL |
| No. per Pack | 1 Each |
| Type | Buret Cylinder |