Learn More
Lenz Laborglasinstrumente™ DURAN™ Borosilicate Glass Round Bottom Flasks with Conical Ground Joint
Brand:  Lenz Laborglasinstrumente™ 03001928
Additional Details : Weight : 0.03700kg
 View more versions of this product
Product Code. 10697281
  • 74.41 EUR / Pack of 10
Estimated Shipment: 30-05-2024
Specifications
According to DIN EN ISO 4797 and in addition to standard
Borosilicate Glass, Duran Glass
Round
50 mL
19/26 NS Socket
51 mm