Learn More
DWK Life Sciences™ DURAN™ Blank for Round Bottom Cylinder, for jacketed vessel, rim cracked-off, without print
Product Code. 10040031 Shop All DWK Life Sciences Products
Click to view available options
Diameter (Metric) Outer:
105 mm
130 mm
160 mm
185 mm
215 mm
250 mm
Unit Size:
Each
Product Code. 10040031
Brand: DWK Life Sciences™ 210275879
Only null left
Add to basket
Add to basket
For processing as a reaction vessel.
DURAN™ cylinders with a round bottom are convincing due to their good wall thickness distribution and enable the cost-effective production of reaction vessels with reproducible geometry. For pressure resistance, please refer to our notes in the technical appendix of the laboratory glassware catalogue. For glass articles which deviate from the original form due to further processing, the permissible operating data must be determined in accordance with the technical rules and regulations.
  • For jacket vessels edge blown off
  • Component for further processing into reaction vessels
  • Cost-effective production of reaction vessels with reproducible geometry
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Description Autoclavable
Diameter (Metric) Outer 185 mm
For Use With (Application) For processing as a reaction vessel
Includes 1/Pk.
Depth (Metric) Interior 4.5 mm wall thickness
Material Borosilicate Glass (3.3)
Product Content Correction
Product Title
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato