Learn More
Corning™ Dulbecco's Phosphate Buffered Saline
BIOPHARMA
Product Code. p-8010455
Click to view available options
Quantity:
10,000 mL
20,000 mL
6 x 500 mL
Concentration:
10X
1X
Unit Size:
10 L
20 L
Pack of 6
Product Code. 15343571
Brand: Corning™ 20031CV
For use alone or combined with other agents for washing tissues and cells
  • Provide cells with water and inorganic ions
  • Maintains physiological pH and osmotic pressure
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material DPBS
Concentration 10X
Quantity 6 x 500 mL