Learn More
IKA™ DS 5 Tube Attachment
Shop All IKA Products
Click to view available options
Holds:
50 mL
Unit Size:
Each
Specifications
Specifications
| Holds | 50 mL |
| Tube Capacity | 50 mL |
| Product Type | DS 5 Tube Attachment |
| Diameter (Metric) | 28 mm |