Learn More
IKA™ DS 3 Tube Attachment
Shop All IKA Products
Click to view available options
Holds:
15 mL
Unit Size:
Each
Specifications
Specifications
| Holds | 15 mL |
| Tube Capacity | 15 mL |
| Product Type | DS 3 Tube Attachment |
| Diameter (Metric) | 16 mm |