missing translation for 'onlineSavingsMsg'
Learn More

Droxinostat, MedChemExpress

Product Code. 16408307
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16408307 5 mg 5mg
16446146 100 mg 100mg
16435608 10 mg 10mg
16421777 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16408307 Supplier Medchem Express Supplier No. HY13267

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Droxinostat(NS41080) is a selective inhibitor of HDAC3, HDAC6, and HDAC8 with IC50 of 16.9, 2.47 and 1.46 μM, respectively; > 8-fold selective against HDAC3 and no inhibition to HDAC1, 2, 4, 5, 7, 9, and 10.

Droxinostat(NS41080) is a selective inhibitor of HDAC3, HDAC6, and HDAC8 with IC50 of 16.9, 2.47 and 1.46 μM, respectively

Specifications

Chemical Name or Material Droxinostat
CAS 99873-43-5
Color White
Purity Grade Notes Research
Molecular Formula C11H14ClNO3
Quantity 5 mg
Synonym NS 41080
Solubility Information DMSO : ≥ 150 mg/mL (615.54 mM)
SMILES ClC1=CC=C(C(C)=C1)OCCCC(NO)=O
Molecular Weight (g/mol) 243.69
Formula Weight 243.69
Percent Purity 98.78%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.