missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Drostanolone Propionate, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Drostanolone Propionate |
| CAS | 521-12-0 |
| Analyte or Component Names | Drostanolone Propionate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H36 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 2MDTP, 32379, Drolban, Dromostanolone propionate, Drostanolone propionate, Emdisterone, MDHT, Masterid, Masteril, Masteron, Masterone, Medrotestron propionate, Medrotestrone propanoate, Medrotestrone propionate, Permastril, Prometholone, (2alpha,5alpha,17beta)-2-Methyl-17-(1-oxopropoxy)-androstan-3-one, 5alpha-Androstan-3-one, 17beta-hydroxy-2alpha-methyl-, propionate (6CI,7CI,8CI), 17beta-Hydroxy-2alpha-methyl-5alpha-androstan-3-one propionate, 17beta-Hydroxy-2alpha-methylandrostan-3-one propionate, 2alpha-Methyl-17beta-hydroxy-5alpha-androstan-3-one 17-propionate, 2alpha-Methyl-17beta-propionoxy-5alpha-androstan-3-one, 2alpha-Methyl-4,5-dihydrotestosterone propionate, 2alpha-Methylandrostan-17beta-ol-3-one propionate, 2alpha-Methyldihydrotestosterone propionate |
| InChI Formula | InChI=1S/C23H36O3/c1-5-21(25)26-20-9-8-17-16-7-6-15-12-19(24)14(2)13-23(15,4)18(16)10-11-22(17,20)3/h14-18,20H,5-13H2,1-4H3/t14-,15+,16+,17+,18+,20+,22+,23+/m1/s1 |
| Solution Type | Neat |
| SMILES | CCC(=O)O[C@H]1CC[C@H]2[C@@H]3CC[C@H]4CC(=O)[C@H](C)C[C@]4(C)[C@H]3CC[C@]12C |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?